missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ IL-33 Antibody (itepekimab) - IgG4SP, Novus Biologicals™
Human Monoclonal Antibody
Brand: Novus Biologicals™ NBP3-28407-1mg
This item is not returnable.
View return policy
Description
IL-33 Monoclonal antibody specifically detects IL-33 in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay
Specifications
| IL-33 | |
| Monoclonal | |
| Unconjugated | |
| O95760 | |
| Human | |
| Protein A purified | |
| RUO | |
| Primary | |
| Reconstitute with sterile, distilled water to a final concentration of 1 mg/mL. Gently shake to solubilize completely. Do not vortex. | |
| Store at -20°C in powder form. Store at -80°C once reconstituted. | |
| IgG4 |
| Flow Cytometry, ELISA, Functional Assay | |
| itepekimab | |
| Lyophilized from 25mM histidine, 8% sucrose, 0.01% Tween80 (pH6.2) | |
| C9orf26chromosome 9 open reading frame 26 (NF-HEV), DKFZp586H0523, DVS27, DVS27-related protein, IL1F11, IL-1F11, IL-33, interleukin 33, Interleukin-1 family member 11, interleukin-33, NFHEV, NF-HEVNFEHEV, Nuclear factor from high endothelial venules, RP11-575C20.2 | |
| IL-33 | |
| 1 mg | |
| Biologically Active Proteins, Cellular Markers, Core ESC Like Genes, Immunology, Inflammation, Innate Immunity, Neuroscience, Sensory Systems, Stem Cell Markers, Stem Cells, Transcription Factors and Regulators, Vision | |
| 90865 | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction