missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD40/TNFRSF5 Antibody (A54 scFv), Janelia Fluor™ 646, Novus Biologicals™
Shop All Bio Techne ProductsDescription
CD40/TNFRSF5 Monoclonal antibody specifically detects CD40/TNFRSF5 in Human samples. It is validated for ELISA, Immunocytochemistry/ Immunofluorescence, Functional Assay, Neutralization
Specifications
Specifications
| Conjugate | Janelia Fluor 646 |
| Dilution | ELISA, Immunocytochemistry/ Immunofluorescence, Functional, Block/Neutralize |
| Gene ID (Entrez) | 958 |
| Form | Purified |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?