missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD40/TNFRSF5 Antibody (5D12), Alexa Fluor™ 532, Novus Biologicals™
Human Monoclonal Antibody
Brand: Novus Biologicals NBP2-81104AF532
This item is not returnable.
View return policy
Description
CD40/TNFRSF5 Monoclonal specifically detects CD40/TNFRSF5 in Human, Cynomolgus Monkey samples. It is validated for Flow Cytometry, ELISA, Immunohistochemistry.
Specifications
| CD40/TNFRSF5 | |
| Monoclonal | |
| Alexa Fluor 532 | |
| 50mM Sodium Borate | |
| CD40 | |
| Anti-CD40/TNFRSF5 mAb 5D12, was generated by immunizing mice with sf9 insect cells expressing recombinant human CD40/TNFRSF5 and selected for the ability to bind EBV-immortalized human B cells. The variable regions from mu5D12 were cloned and used to construct chimeric humanized IgG4 5D12. | |
| 0.1 mL | |
| Adaptive Immunity, Apoptosis, Asthma, B Cell Development and Differentiation Markers, Cancer, Hematopoietic Stem Cell Markers, Immunology, Signal Transduction, Stem Cell Markers | |
| 958 | |
| Human, Cynomolgus Monkey | |
| Purified |
| Flow Cytometry, ELISA, Immunohistochemistry | |
| 5D12 | |
| Flow Cytometry, ELISA, Immunohistochemistry | |
| B cell surface antigen CD40, B-cell surface antigen CD40, Bp50B cell-associated molecule, CD40 antigen, CD40 molecule, TNF receptor superfamily member 5, CD40 type II isoform, CD40L receptor, CDw40, MGC9013, nerve growth factor receptor-related B-lymphocyte activation molecule, p50, TNFRSF5CD40 antigen (TNF receptor superfamily member 5), tumor necrosis factor receptor superfamily member 5, tumor necrosis factor receptor superfamily, member 5 | |
| Human | |
| Protein A purified | |
| RUO | |
| Primary | |
| ch5D12 binds specifically to recombinant and native human CD40/TNFRSF5 ectodomain and has been shown to also bind CD40/TNFRSF5 in cynomolgus and marmoset monkeys. The antibody requires residues within D1 and D1/B2 for binding (Hager et al, 2003) (D1 domain is crucial for CD40L binding). Binding characteristics are very similar between humanized ch5D12 and mu5D12. CD40/TNFRSF5 is a glycoprotein of the TNFR superfamily and is expressed on all mature B cells, dendritic cells, activated monocytes, some endothelial cells and some epithelium including the thymus. CD40/TNFRSF5 binds to CD40L, and this interaction is involved in B-cell activation and proliferation, antigen-presenting cell (APC) activation, initiation of antigen-specific T-cell responses, immunoglobulin production, activation of effector macrophages isotype switching, homotypic adhesion and rescue from apoptosis. | |
| Store at 4°C in the dark. | |
| IgG4 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction