missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD40 Mouse anti-Human, Clone: C40/1605, Abnova™
Mouse Monoclonal Antibody
Brand: Abnova MAB14706.100ug
This item is not returnable.
View return policy
Description
Mouse monoclonal antibody raised against full length recombinant human CD40.
- High specificity
- High reactivity
Specifications
| CD40 | |
| Monoclonal | |
| Unconjugated | |
| Flow Cytometry (0.5-1 ug/million cells) Immunofluorescence (1-2 ug/mL) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (1-2 ug/mL) Western Blot (0.5-1 ug/mL) The optimal working dilution should be determined by the end use | |
| Bp50/CDW40/MGC9013/TNFRSF5/p50 | |
| Mouse | |
| Protein A/G purification | |
| RUO | |
| 958 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG1 κ |
| Flow Cytometry, Immunofluorescence, Immunohistochemistry (PFA fixed), Western Blot | |
| C40/1605 | |
| Mouse monoclonal antibody raised against full length recombinant human CD40. | |
| In 10 mM PBS. | |
| CD40 | |
| Recombinant protein corresponding to full-length human CD40. | |
| 100 μg | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction