missing translation for 'onlineSavingsMsg'
Learn More

CD40 Ligand/TNFSF5 Antibody (hu5c8 (Ruplizumab)), Alexa Fluor™ 594, Novus Biologicals™

Product Code. 18369550 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18369550 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18369550 Supplier Novus Biologicals Supplier No. NBP275907AF594

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Human Monoclonal Antibody

CD40 Ligand/TNFSF5 Monoclonal specifically detects CD40 Ligand/TNFSF5 in Human samples. It is validated for Western Blot, Flow Cytometry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD40 Ligand/TNFSF5
Applications Western Blot, Flow Cytometry
Classification Monoclonal
Clone hu5c8 (Ruplizumab)
Conjugate Alexa Fluor 594
Dilution Western Blot, Flow Cytometry
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias CD154, CD154 antigen, CD40 antigen ligand, CD40 ligand, CD40-L, CD40LIGM, gp39, hCD40L, HIGM1, T-B cell-activating molecule, T-BAM, T-cell antigen Gp39, TNF-related activation protein, TNFSF5IMD3, TRAPtumor necrosis factor (ligand) superfamily, member 5 (hyper-IgM syndrome), tumor necrosis factor (ligand) superfamily member 5, Tumor necrosis factor ligand superfamily member 5
Gene Symbols CD40LG
Host Species Human
Immunogen Human CD40L
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 959
Test Specificity The antibody binds to CD40L with an IC50 of 0.845 ug/ml.
Target Species Human
Content And Storage Store at 4°C in the dark.
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.