missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD4 Rat, PerCP, Clone: RM4-5, enQuireBio™
Rat Monoclonal Antibody
£165.90 - £342.30
Specifications
| Antigen | CD4 |
|---|---|
| Clone | RM4-5 |
| Concentration | 0.5mg/mL |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
15922618
|
EnquireBio
QAB9-PCP-25ug |
25 μg |
£165.90
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
15912618
|
EnquireBio
QAB9-PCP-100ug |
100 μg |
£342.30
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Specifications
| CD4 | |
| 0.5mg/mL | |
| Monoclonal | |
| Rat | |
| 10 mM NaH2PO4, 150 mM NaCl, 0.09% NaN3, 0.1% gelatin, pH7.2 | |
| 12504 | |
| IgG2a κ | |
| Protein A/G Purified |
| RM4-5 | |
| Flow Cytometry | |
| PerCP | |
| RUO | |
| NP_038516 | |
| Cd4 | |
| Primary | |
| Store at 2-4°C. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title