Learn More
CD4 Rat, PE-Cy5, Clone: GK1.5, enQuireBio™
Rat Monoclonal Antibody
Brand:  EnquireBio QAB8-PE5-25ug
 View more versions of this product
Product Code. 15982518
  • £95.92 / 25µg
Specifications
CD4
Monoclonal
0.5mg/mL
10 mM NaH2PO4, 150 mM NaCl, 0.09% NaN3, pH7.2
Cd4
Protein A/G Purified
RUO
12504
IgG2b κ
Flow Cytometry
GK1.5
PE-Cyanine5
NP_038516
Rat
25 μg
Primary
Store at 2-4°C.