missing translation for 'onlineSavingsMsg'
Learn More

CD4 Rat, PE, Clone: RM4-5, enQuireBio™

Rat Monoclonal Antibody

Brand:  EnquireBio QAB9-PE-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15952618

  • £71.60 / 100µg
Estimated Shipment: 20-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

CD4
Monoclonal
0.5mg/mL
10 mM NaH2PO4, 150 mM NaCl, 0.09% NaN3, 0.1% gelatin, pH7.2
Cd4
Protein A/G Purified
RUO
12504
IgG2a κ
Flow Cytometry
RM4-5
PE
NP_038516
Rat
100 μg
Primary
Store at 2-4°C.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers