missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD4 Rat, anti-Mouse, PE, Clone: RM4-5, STEMCELL Technologies
Rat Monoclonal Antibody
POA - £226.80
Specifications
| Antigen | CD4 |
|---|---|
| Clone | RM4-5 |
| Applications | Immunohistochemistry, Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence |
| Classification | Monoclonal |
| Conjugate | PE |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
17178481
|
Stemcell Technologies
60017PE.1 |
50 μg | |||||||
17168481
|
Stemcell Technologies
60017PE |
200 μg |
£226.80
200µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
- The RM4-5 antibody reacts with mouse CD4, an ∼55 kDa single-chain type 1 transmembrane glycoprotein and member of the immunoglobulin (Ig) superfamily; CD4 contains four extracellular Ig-like domains
- CD4 is expressed at relatively high levels by most thymocytes and a subpopulation of T cells (T helper/inducer cells), and at lower levels on dendritic cells
- CD4 binds to a non-polymorphic region of MHC II and acts as a co-receptor to the T cell receptor (TCR) in MHC II-restricted antigen recognition by enhancing the avidity of the association between the TCR and MHC II-antigen complex
- Binding of the RM4-5 antibody to CD4 inhibits ligand binding in vitro; binding of the RM4-5 antibody can be blocked by the clone GK1.5 antibody
- This antibody clone has been verified for purity assessments of cells isolated with EasySep™ kits, including EasySep™ Mouse CD4+ T Cell Isolation Kit (Catalog #19852) and EasySep™ Mouse CD25 Regulatory T Cell Positive Selection Kit (Catalog #18782)
Specifications
| CD4 | |
| Immunohistochemistry, Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence | |
| PE | |
| Mouse | |
| Primary |
| RM4-5 | |
| Monoclonal | |
| Rat | |
| IgG2a κ |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title