Learn More
CD4 Rat anti-Mouse, CoraLite 647, Clone: GK1.5, Proteintech
Rat Monoclonal Antibody
Brand: Proteintech CL647-65104-100TESTS
Additional Details : Weight : 0.00010kg
Description
The CD4 antibody from Proteintech is a rat monoclonal antibody generated with mouse ctl clone v4 of mouse CD4. This antibody recognizes mouse antigen. The CD4 antibody has been validated for the following applications: IF, FC analysis. This antibody is coralite™647.
Specifications
CD4 | |
Monoclonal | |
5 μL/Test | |
Rat | |
Primary | |
Mouse |
Immunofluorescence | |
GK1.5 | |
CoraLite 647 | |
100 Tests | |
12504 | |
IgG2b |