Learn More
CD4 Mouse anti-Human, PE, Clone: RPA-T4, BD
Mouse Monoclonal Antibody
£98.44 - £969.55
Specifications
| Antigen | CD4 |
|---|---|
| Clone | RPA-T4 |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Conjugate | PE |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
15881389
|
BD Biosciences
561843 |
25 Tests |
£102.00 £98.44 / Each Save £3.56 3% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
15818088
|
BD Biosciences
555347 |
100 Tests |
£180.00
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
15891389
|
BD Biosciences
561844 |
500 Tests |
£1010.00 £969.55 / Each Save £40.45 4% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The RPA-T4 monoclonal antibody specifically binds to CD4, a 59 kDa single-chain transmembrane glycoprotein that is expressed on T-helper/inducer cell populations. CD4 is also expressed on thymocytes subsets and at lower levels on monocytes and macrophages. CD4 functions as a co-receptor in MHC class II-restricted antigen-induced T cell activation and as a receptor for human immunodeficiency viruses (HIV). This antibody binds to the D1 domain (CDR1 and CDR3 epitopes) of the CD4 antigen and reacts with approximately 80% of thymocytes and 45% of peripheral blood lymphocytes. RPA-T4 is capable of blocking HIV-1, gp120, and inhibits syncytium formation.
This antibody is routinely tested by flow cytometric analysis. Other applications were tested at BD Biosciences Pharmingen during antibody development or are reported in the literature.
Specifications
| CD4 | |
| Flow Cytometry | |
| PE | |
| RUO | |
| Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
| RPA-T4 | |
| Monoclonal | |
| Mouse | |
| Human | |
| IgG1 κ | |
| Affinity Purified |
For Research Use Only.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.