missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD4 Mouse anti-Human , BV421, Clone: OKT4, BD Biosciences™
Mouse Monoclonal Antibody
£170.62 - £325.79
Specifications
| Antigen | CD4 |
|---|---|
| Clone | OKT4 |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Conjugate | Brilliant Violet 421 |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
17227544
|
BD Biosciences
566706 |
25 Tests |
£173.00 £170.62 / 25 tests Save £2.38 1% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
17217544
|
BD Biosciences
566703 |
100 Tests |
£333.00 £325.79 / 100 tests Save £7.21 2% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CD4 Monoclonal antibody specifically detects CD4 in Human samples. It is validated for Flow CytometrySpecifications
| CD4 | |
| Flow Cytometry | |
| Brilliant Violet 421 | |
| RUO | |
| Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
| Sheep Erythrocyte Rosette-purified Human T Cells | |
| Primary |
| OKT4 | |
| Monoclonal | |
| Mouse | |
| Human | |
| CD4 antigen (p55); T-cell surface antigen T4/Leu-3 | |
| IgG2b κ | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated with BD Horizon BV421 under optimum conditions, and unconjugated antibody and free BD Horizon BV421 were removed. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title