missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD4 Mouse anti-Human, BUV805, Clone: SK3, BD Horizon™
Mouse Monoclonal Antibody
Brand: BD Biosciences 564911
This item is not returnable.
View return policy
Description
CD4 Monoclonal antibody specifically detects CD4 in Human samples. It is validated for Flow cytometry.
CD4 Monoclonal antibody specifically detects CD4 in Human samples. It is validated for Flow cytometry.
Specifications
| CD4 | |
| Monoclonal | |
| Brilliant Ultraviolet 805 | |
| Aqueous buffered solution containing ≤0.09% sodium azide. | |
| Mouse | |
| 25 Tests | |
| Cell Biology | |
| Human | |
| Purified |
| Flow Cytometry | |
| SK3 | |
| Vol. Per test: 5μL | |
| L3T4; Leu3a; T-cell surface antigen T4/Leu-3 ; W3/25 ; CD4 antigen (p55) | |
| Human Peripheral Blood T Cells | |
| RUO | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated with BD Horizon BUV805 under optimum conditions and unconjugated antibody and free BD Horizon BUV805 were removed. | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only.
Spot an opportunity for improvement?Share a Content Correction