Learn More
CD4 Mouse anti-Human, Brilliant Ultraviolet 737, Clone: SK3 (also known as Leu3a), BD Horizon™
Mouse Monoclonal Antibody
£197.74 - £463.10
Specifications
| Antigen | CD4 |
|---|---|
| Clone | SK3 (also known as Leu3a) |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Conjugate | Brilliant Ultraviolet 737 |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
17237854
|
BD Biosciences
612749 |
25 Tests |
£202.00 £197.74 / 25 tests Save £4.26 2% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
17227854
|
BD Biosciences
612748 |
100 Tests |
£472.00 £463.10 / 100 tests Save £8.90 1% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The SK3 monoclonal antibody specifically binds to CD4. The CD4 antigen is a 55 kDa type 1 transmembrane glycoprotein that belongs to the immunoglobulin superfamily. It is expressed on T-helper/inducer lymphocytes and monocytes. CD3+CD4+ T cells comprise approximately 28% to 58% of normal peripheral blood lymphocytes. CD4 is also expressed on 80% to 95% of normal thymocytes. The CD4 antigen is present in low density on the cell surface of monocytes and in the cytoplasm of monocytes and macrophages (CD3-CD4+). CD4 serves as a T cell coreceptor for MHC class II antigen recognition and as a receptor for the human immunodeficiency virus (HIV). The SK3 antibody inhibits HIV binding to CD4+ cells.
The antibody was conjugated to BD Horizon™ BUV737 which is part of the BD Horizon Brilliant™ Ultraviolet family of dyes. This dye is a tandem fluorochrome of BD Horizon BUV395 with an Ex Max of 348-nm and an acceptor dye with an Em Max at 737-nm. BD Horizon Brilliant BUV737 can be excited by the ultraviolet laser (355 nm) and detected with a 740/35 filter. Due to the excitation of the acceptor dye by other laser lines, there may be significant spillover into channels detecting Alexa Fluor™ 700-like dyes (e.g., 712/20-nm filter).
Specifications
| CD4 | |
| Flow Cytometry | |
| Brilliant Ultraviolet 737 | |
| RUO | |
| Aqueous buffered solution containing ≤0.09% sodium azide. | |
| IgG1 κ | |
| Affinity Purified |
| SK3 (also known as Leu3a) | |
| Monoclonal | |
| Mouse | |
| Human | |
| Human Peripheral Blood T Cells | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.