missing translation for 'onlineSavingsMsg'
Learn More

CD4 Mouse anti-Human, Alexa Fluor 700, Clone: RPA-T4, BD

Mouse Monoclonal Antibody

£161.58 - £205.27

Specifications

Antigen CD4
Clone RPA-T4
Concentration 0.2mg/mL
Applications Flow Cytometry
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15880699
View Documents
BD Biosciences
561030
25 μg
ONLINE EXCLUSIVE PRICE
£168.00
£161.58 / Each
Save £6.42
3% Off
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
15899108
View Documents
BD Biosciences
557922
0.1 mg
ONLINE EXCLUSIVE PRICE
£213.00
£205.27 / Each
Save £7.73
3% Off
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

The RPA-T4 monoclonal antibody specifically binds to CD4, a 59 kDa single-chain transmembrane glycoprotein that is expressed on T-helper/inducer cell populations. CD4 is also expressed on thymocytes subsets and at lower levels on monocytes and macrophages. CD4 functions as a co-receptor in MHC class II-restricted antigen-induced T cell activation and as a receptor for human immunodeficiency viruses (HIV). This antibody binds to the D1 domain (CDR1 and CDR3 epitopes) of the CD4 antigen and reacts with approximately 80% of thymocytes and 45% of peripheral blood lymphocytes. RPA-T4 is capable of blocking HIV-1, gp120, and inhibits syncytium formation.

This antibody is routinely tested by flow cytometric analysis. Other applications were tested at BD Biosciences Pharmingen during antibody development only or reported in the literature.

Specifications

Specifications

CD4
0.2mg/mL
Monoclonal
Mouse
Human
IgG1 κ
Affinity Purified
RPA-T4
Flow Cytometry
Alexa Fluor 700
RUO
Aqueous buffered solution containing protein stabilizer and ≤0.09% sodium azide.
Primary
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.