missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CD4 Monoclonal Antibody (W3/25), FITC

Mouse Monoclonal Antibody
£233.00 - £462.00
Specifications
| Antigen | CD4 |
|---|---|
| Clone | W3/25 |
| Concentration | 0.1 mg/mL |
| Content And Storage | Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. Store in the dark. |
| Applications | Flow Cytometry, Immunocytochemistry |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15388290
![]() |
Invitrogen™
MA517390 |
100 μg |
£233.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
15398290
|
Invitrogen™
MA517391 |
500 μg |
£462.00
500µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CD4 Monoclonal Antibody for ICC/IF, Flow
The CD4 antigen is involved in the recognition of MHC class II molecules and is a co-receptor for HIV. CD4 is primarily expressed in a subset of T-lymphocytes, also referred to as T helper cells, but may also be expressed by other cells in the immune system, such as monocytes, macrophages, and dendritic cells. At the tissue level, CD4 expression may be detected in thymus, lymph nodes, tonsils, and spleen, and also in specific regions of the brain, gut, and other non-lymphoid tissues. CD4 functions to initiate or augment the early phase of T-cell activation through its association with the T-cell receptor complex and protein tyrosine kinase, Lck. It may also function as an important mediator of direct neuronal damage in infectious and immune-mediated diseases of the central nervous system. Multiple alternatively spliced transcripts have been identified in this gene [RefSeq, July 2017].Specifications
| CD4 | |
| 0.1 mg/mL | |
| Flow Cytometry, Immunocytochemistry | |
| FITC | |
| Mouse | |
| RUO | |
| PBS with 4-5mg/mL BSA and 0.02% sodium azide | |
| P05540 | |
| 24932 | |
| Rat thymocyte membrane. | |
| Antibody |
| W3/25 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. Store in the dark. | |
| Monoclonal | |
| Liquid | |
| IgG1 | |
| Rat | |
| CD4 | |
| Activation B7-1 antigen; B7; B7.1; B7-1; BB1; B-lymphocyte activation antigen B7; CD28LG; CD28LG1; CD4; CD4 antigen; CD4 antigen (p55); CD4 antigen p55; Cd4 molecule; CD4 precursor; CD4 receptor; CD4, allele 1; cd4a; CD4mut; CD80; CD80 antigen (CD28 antigen ligand 1, B7-1 antigen); CD80 molecule; cell surface glycoprotein CD4; costimulatory factor CD80; costimulatory molecule variant IgV-CD80; CTLA-4 counter-receptor B7.1; fCD4; L3T4; LAB7; Leu-3; Ly-4; lymphocyte antigen CD4; lymphocyte antigen CD4 precursor; membrane protein; p55; T-cell differentiation antigen L3T4; T-cell surface antigen T4/Leu-3; T-cell surface glycoprotein CD4; T-cell surface glycoprotein CD4 precursor (T-cell surface antigen T4/Leu-3) (T-cell differentiation antigen L3T4); T-lymphocyte activation antigen CD80; W3/25; W3/25 antigen | |
| CD4 | |
| Primary | |
| Protein G |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title