missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CD4 Monoclonal Antibody (SK3 (SK-3)), NovaFluor™ Red 755, eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
£157.00 - £346.00
Specifications
| Antigen | CD4 |
|---|---|
| Clone | SK3 (SK-3) |
| Concentration | 0.1 μg/Test |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30055247
|
Invitrogen™
H001T03R06-A |
100 Tests |
£346.00
Pack of 100 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30055597
|
Invitrogen™
H001T02R06-A |
25 Tests |
£157.00
Pack of 25 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CD4 Monoclonal antibody specifically detects CD4 in Human samples. It is validated for Flow CytometrySpecifications
| CD4 | |
| 0.1 μg/Test | |
| Monoclonal | |
| Mouse | |
| Human | |
| P01730 | |
| 920 | |
| Primary | |
| 4°C, store in dark, DO NOT FREEZE! |
| SK3 (SK-3) | |
| Flow Cytometry | |
| NovaFluor Red 755 | |
| RUO | |
| PBS with 0.09% sodium azide; pH 7.2 | |
| Activation B7-1 antigen; B7; B7.1; B7-1; BB1; B-lymphocyte activation antigen B7; CD28LG; CD28LG1; CD4; CD4 antigen; CD4 antigen (p55); CD4 antigen p55; Cd4 molecule; CD4 precursor; CD4 receptor; CD4, allele 1; cd4a; CD4mut; CD80; CD80 antigen (CD28 antigen ligand 1, B7-1 antigen); CD80 molecule; cell surface glycoprotein CD4; costimulatory factor CD80; costimulatory molecule variant IgV-CD80; CTLA-4 counter-receptor B7.1; fCD4; L3T4; LAB7; Leu-3; Ly-4; lymphocyte antigen CD4; lymphocyte antigen CD4 precursor; membrane protein; p55; T-cell differentiation antigen L3T4; T-cell surface antigen T4/Leu-3; T-cell surface glycoprotein CD4; T-cell surface glycoprotein CD4 precursor (T-cell surface antigen T4/Leu-3) (T-cell differentiation antigen L3T4); T-lymphocyte activation antigen CD80; W3/25; W3/25 antigen | |
| IgG1 κ | |
| Affinity chromatography | |
| CD4 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title