Learn More
Invitrogen™ CD4 Monoclonal Antibody (S3.5), PE

Mouse Monoclonal Antibody
£253.00 - £764.00
Specifications
| Antigen | CD4 |
|---|---|
| Clone | S3.5 |
| Content And Storage | 4°C, store in dark |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10680664
![]() |
Invitrogen™
MHCD0404 |
500 μL |
£253.00
500µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
10775133
|
Invitrogen™
MHCD04044 |
2 mL |
£764.00
2mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
R-phycoerythrin (PE) is a stable and highly soluble phycobiliprotein which provides maximal absorbance and fluorescence without susceptibility to internal or external fluorescence quenching, thus providing an exceptional quantum yields and molar extinction coefficients. This antibody may be used, in combination with other indicators, for the diagnosis or prognosis of immunodeficiency diseases, including hypogammaglobulinemia, severe combined immunodeficiency (SCID), and the acquired immunodeficiency disease (AIDS).
The CD4 antigen is involved in the recognition of MHC class II molecules and is a co-receptor for HIV. CD4 is primarily expressed in a subset of T-lymphocytes, also referred to as T helper cells, but may also be expressed by other cells in the immune system, such as monocytes, macrophages, and dendritic cells. At the tissue level, CD4 expression may be detected in thymus, lymph nodes, tonsils, and spleen, and also in specific regions of the brain, gut, and other non-lymphoid tissues. CD4 functions to initiate or augment the early phase of T-cell activation through its association with the T-cell receptor complex and protein tyrosine kinase, Lck. It may also function as an important mediator of direct neuronal damage in infectious and immune-mediated diseases of the central nervous system. Multiple alternatively spliced transcripts have been identified in this gene [RefSeq, July 2017].Specifications
| CD4 | |
| 4°C, store in dark | |
| Monoclonal | |
| Liquid | |
| IgG2a | |
| Human | |
| CD4 | |
| Activation B7-1 antigen; B7; B7.1; B7-1; BB1; B-lymphocyte activation antigen B7; CD28LG; CD28LG1; CD4; CD4 antigen; CD4 antigen (p55); CD4 antigen p55; Cd4 molecule; CD4 precursor; CD4 receptor; CD4, allele 1; cd4a; CD4mut; CD80; CD80 antigen (CD28 antigen ligand 1, B7-1 antigen); CD80 molecule; cell surface glycoprotein CD4; costimulatory factor CD80; costimulatory molecule variant IgV-CD80; CTLA-4 counter-receptor B7.1; fCD4; L3T4; LAB7; Leu-3; Ly-4; lymphocyte antigen CD4; lymphocyte antigen CD4 precursor; membrane protein; p55; T-cell differentiation antigen L3T4; T-cell surface antigen T4/Leu-3; T-cell surface glycoprotein CD4; T-cell surface glycoprotein CD4 precursor (T-cell surface antigen T4/Leu-3) (T-cell differentiation antigen L3T4); T-lymphocyte activation antigen CD80; W3/25; W3/25 antigen | |
| CD4 | |
| Primary | |
| Purified |
| S3.5 | |
| Flow Cytometry | |
| PE | |
| Mouse | |
| RUO | |
| PBS with BSA, sucrose and 0.1% sodium azide | |
| P01730 | |
| 920 | |
| Human CD4. | |
| Antibody |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.