Learn More
CD4 Monoclonal Antibody (RM4-5), eBioscience™, Invitrogen™

Rat Monoclonal Antibody
£31.79 - £290.00
Specifications
| Antigen | CD4 |
|---|---|
| Clone | RM4-5 |
| Concentration | 0.5 mg/mL |
| Applications | Flow Cytometry, Functional Assay, Immunohistochemistry (Frozen), Neutralization |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
15296747
|
Invitrogen
14-0042-81 |
50 μg | |||||||
15206757
![]() |
Invitrogen™
14-0042-82 |
100 μg |
£73.75
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
15216757
|
Invitrogen™
14-0042-85 |
500 μg |
£161.00
500µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
15226757
|
Invitrogen™
14-0042-86 |
1 mg |
£290.00
1mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Description: The RM4-5 monoclonal antibody reacts with the mouse CD4 molecule, a 55 kDa cell surface receptor expressed by a majority of thymocytes, subpopulation of mature T cells and dendritic cells. CD4 binds to MHC class II on the surface of antigen presenting cells and plays an important role both in T cell development and in optimal functioning of mature T cells. In T cells, CD4 associates with protein tyrosine kinase p56lck through its cytoplasmic tail. Binding of RM4-5 is blocked by GK1.5. Applications Reported: The RM4-5 antibody has been reported for use in flow cytometric analysis, and immunohistochemical staining. It has also been reported in isolation and depletion of CD4+ cells and blocking of ligand binding. (Please use Functional Grade purified RM4-5, cat. 16-0042, in functional assays.). Applications Tested: The RM4-5 antibody has been tested by flow cytometric analysis of mouse thymocytes and splenocytes. This can be used at less than or equal to 0.5 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Purity: Greater than 90%, as determined by SDS-PAGE. Aggregation: Less than 10%, as determined by HPLC. Filtration: 0.2 μm post-manufacturing filtered.
The CD4 antigen is involved in the recognition of MHC class II molecules and is a co-receptor for HIV. CD4 is primarily expressed in a subset of T-lymphocytes, also referred to as T helper cells, but may also be expressed by other cells in the immune system, such as monocytes, macrophages, and dendritic cells. At the tissue level, CD4 expression may be detected in thymus, lymph nodes, tonsils, and spleen, and also in specific regions of the brain, gut, and other non-lymphoid tissues. CD4 functions to initiate or augment the early phase of T-cell activation through its association with the T-cell receptor complex and protein tyrosine kinase, Lck. It may also function as an important mediator of direct neuronal damage in infectious and immune-mediated diseases of the central nervous system. Multiple alternatively spliced transcripts have been identified in this gene [RefSeq, July 2017].Specifications
| CD4 | |
| 0.5 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| P06332 | |
| IgG2a κ | |
| Affinity chromatography | |
| Antibody |
| RM4-5 | |
| Flow Cytometry, Functional Assay, Immunohistochemistry (Frozen), Neutralization | |
| Unconjugated | |
| Rat | |
| Mouse | |
| 12504 | |
| Primary | |
| 4° C |
For Research Use Only.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.