missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ CD4 Monoclonal Antibody (OX35), FITC, eBioscience™, Invitrogen™

Product Code. 15237989
Change view
Click to view available options
Quantity:
100 μg
50 μg
500 μg
Unit Size:
100µg
500µg
50µg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15237989 50 μg 50µg
15247989 100 μg 100µg
15257989 500 μg 500µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15237989 Supplier Invitrogen™ Supplier No. 11004081

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Description: The OX35 monoclonal antibody reacts with rat CD4. CD4 is a co-receptor which participates in signaling through the T cell receptor by making contacts with MHC class II expressed on antigen-presenting cells (APC). CD4 is expressed in the thymus by CD4+CD8+ double-positive and CD4+ single-positive thymocytes, and by CD4+ helper T cells in the periphery. CD4 is a cell-surface membrane glycoprotein which contains four extracellular immunoglobulin-like domains. Applications Reported: This OX35 antibody has been reported for use in flow cytometric analysis. Applications Tested: This OX35 antibody has been tested by flow cytometric analysis of rat splenocytes. This can be used at less than or equal to 0.25 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Excitation: 488 nm; Emission: 520 nm; Laser: Blue Laser. Filtration: 0.2 μm post-manufacturing filtered.

The CD4 antigen is involved in the recognition of MHC class II molecules and is a co-receptor for HIV. CD4 is primarily expressed in a subset of T-lymphocytes, also referred to as T helper cells, but may also be expressed by other cells in the immune system, such as monocytes, macrophages, and dendritic cells. At the tissue level, CD4 expression may be detected in thymus, lymph nodes, tonsils, and spleen, and also in specific regions of the brain, gut, and other non-lymphoid tissues. CD4 functions to initiate or augment the early phase of T-cell activation through its association with the T-cell receptor complex and protein tyrosine kinase, Lck. It may also function as an important mediator of direct neuronal damage in infectious and immune-mediated diseases of the central nervous system. Multiple alternatively spliced transcripts have been identified in this gene [RefSeq, July 2017].
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD4
Applications Flow Cytometry
Classification Monoclonal
Clone OX35
Concentration 0.5 mg/mL
Conjugate FITC
Formulation PBS with 0.09% sodium azide; pH 7.2
Gene CD4
Gene Accession No. P05540
Gene Alias Activation B7-1 antigen; B7; B7.1; B7-1; BB1; B-lymphocyte activation antigen B7; CD28LG; CD28LG1; CD4; CD4 antigen; CD4 antigen (p55); CD4 antigen p55; Cd4 molecule; CD4 precursor; CD4 receptor; CD4, allele 1; cd4a; CD4mut; CD80; CD80 antigen (CD28 antigen ligand 1, B7-1 antigen); CD80 molecule; cell surface glycoprotein CD4; costimulatory factor CD80; costimulatory molecule variant IgV-CD80; CTLA-4 counter-receptor B7.1; fCD4; L3T4; LAB7; Leu-3; Ly-4; lymphocyte antigen CD4; lymphocyte antigen CD4 precursor; membrane protein; p55; T-cell differentiation antigen L3T4; T-cell surface antigen T4/Leu-3; T-cell surface glycoprotein CD4; T-cell surface glycoprotein CD4 precursor (T-cell surface antigen T4/Leu-3) (T-cell differentiation antigen L3T4); T-lymphocyte activation antigen CD80; W3/25; W3/25 antigen
Gene Symbols CD4
Host Species Mouse
Purification Method Affinity chromatography
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 24932
Target Species Rat
Content And Storage 4°C, store in dark, DO NOT FREEZE!
Product Type Antibody
Form Liquid
Isotype IgG2a κ
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.