missing translation for 'onlineSavingsMsg'
Learn More

IBA Lifesciences CD4 Fab-TACS™ Gravity Kit, Mouse

Product Code. 16222931 Shop All IBA Lifesciences Products
Click to view available options
Quantity:
10 Columns
4 Columns
Unit Size:
10 sets
4 sets
This item is not returnable. View return policy

Product Code. 16222931

Brand: IBA Lifesciences 63401004

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

For manual positive cell separation. The Fab-TACS™ Gravity Kit provides label-free target cells without the use of high affinity antibodies and magnetic beads

  • The kit allows the selection of CD4+ T cells from mouse spleen.
  • Fab-TACS™ Gravity column (Capacity: 1x108target cells each)
  • CD4 Fab-Strep, mouse
  • Buffer CI (10x)
  • Biotin stock solution
  • Fab-TACS™ Gravity Adapter
  • The Fab-TACS™ Gravity column is filled with a Strep-Tactin™ coated matrix made of cell-grade agarose.
  • Strep-tagged Fab fragments (Fab-Streps) specifically bind to the matrix.
  • Subsequently, single-cell tissue suspensions pass through the column.
  • Target cells adhere to the matrix based on the exclusive binding of the Fab-Strep to the target cell.
  • Non-target cells are efficiently washed away.
  • In a final step, the addition of biotin causes the elution of the target cells and the Fab-Streps.
  • After elution, the Fab-Streps self-dissociate from the cell surfaces.
  • The label-free authentic target cells are now ready for further downstream applications.
  • Specifications

    Content And Storage 2°C to 8°C
    Quantity 4 Columns
    Type Non-magnetic Cell Isolation Kit
    Species Mouse
    Sufficient For 1x108 target cells/column
    Product Content Correction

    Your input is important to us. Please complete this form to provide feedback related to the content on this product.

    Product Title

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

    Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.