missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
CD4 Monoclonal specifically detects CD4 in Mouse samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Frozen, Flow (Cell Surface), Cell depletion, Inhibition Activity, CyTOF-ready.
Specifications
Specifications
| Antigen | CD4 |
| Classification | Monoclonal |
| Clone | GK1.5 |
| Concentration | 0.5mg/mL |
| Conjugate | Unconjugated |
| Dilution | Flow Cytometry 2.5 ug 1 million cells, Immunohistochemistry 1-25 ug/ml, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunoprecipitation, Immunohistochemistry-Frozen, Flow (Cell Surface) 2.5 ug 1 million cells, Cell depletion, Inhibition Activity, CyTOF-ready |
| Gene Accession No. | NP_000607 |
| Gene Alias | CD4 antigen, CD4 antigen (p55), CD4 molecule, CD4 receptor, CD4mut, T-cell surface antigen T4/Leu-3, T-cell surface glycoprotein CD4 |
| Gene Symbols | CD4 |
| Host Species | Rat |
| Show More |
For Research Use Only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?