missing translation for 'onlineSavingsMsg'
Learn More

CD4 Antibody (CE9.1 (Clenoliximab)) - Azide and BSA Free, Novus Biologicals™

Product Code. 18396350 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.025 mg
Unit Size:
25µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18396350 0.025 mg 25µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18396350 Supplier Novus Biologicals Supplier No. NBP275910SS

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Human Monoclonal Antibody

CD4 Monoclonal specifically detects CD4 in Human, Chimpanzee samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Surface Plasmon Resonance, Block/Neutralize.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD4
Applications Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Blocking Assay, Neutralization
Classification Monoclonal
Clone CE9.1 (Clenoliximab)
Conjugate Unconjugated
Dilution Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Surface Plasmon Resonance, Block/Neutralize
Formulation PBS with 0.02% Proclin 300
Gene Alias biosimilar, biosimilars, CD4 antigen, CD4 antigen (p55), CD4 molecule, CD4 receptor, CD4mut, T-cell surface antigen T4/Leu-3, T-cell surface glycoprotein CD4
Gene Symbols CD4
Host Species Human
Immunogen Domain 1 of human CD4.
Purification Method Protein A purified
Quantity 0.025 mg
Research Discipline Adaptive Immunity, Cell Biology, Cellular Markers, Cytokine Research, Immunology, Innate Immunity, Regulatory Immunology, Signal Transduction, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 920.0
Test Specificity The antibody binds to human CD4 with a Kd of 1.0 nM and an ED50 of 0.2 ug/ml.
Target Species Human, Primate
Content And Storage Store at 4°C for up to 3 months. For longer storage, aliquot and store at -20°C.
Form Purified
Isotype IgG4 λ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.