missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD4 Antibody - BSA Free, Novus Biologicals™
CD4 (cluster of differentiation 4), also known as L3T4 or T4, is a 55 kDa single chain type I transmembrane glycoprotein belonging to the immunoglobin (Ig) superfamily.
Brand: Novus Biologicals NBP1-19371
This item is not returnable.
View return policy
Description
CD4 Polyclonal specifically detects CD4 in Human, Mouse, Rat, Porcine, Canine, Rabbit samples. It is validated for Western Blot, Simple Western, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Dual RNAscope ISH-IHC.
Specifications
| CD4 | |
| Polyclonal | |
| Unconjugated | |
| PBS with 0.02% Sodium Azide | |
| CD4 antigen, CD4 antigen (p55), CD4 molecule, CD4 receptor, CD4mut, T-cell surface antigen T4/Leu-3, T-cell surface glycoprotein CD4 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Rabbit reactivity reported in the scientific literature (PMID: 2212730). Customer feedback positive on canine. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| 1.0 mg/mL | |
| Western Blot 1 ug/ml, Simple Western 1:100, Flow Cytometry 5 ug/ml, Immunohistochemistry 1:50-1:500, Immunocytochemistry/Immunofluorescence 1:50-1:200, Immunohistochemistry-Paraffin 1:50-1:500, Immunohistochemistry-Frozen, Dual RNAscope ISH-IHC | |
| P01730, P06332 | |
| CD4 | |
| A synthetic peptide made to an C-terminal region of the human CD4 Antibody protein (within residues 400-458). [Swiss-Prot P01730] | |
| 0.1 mL | |
| Adaptive Immunity, Cell Biology, Cellular Markers, Cytokine Research, Immunology, Innate Immunity, Regulatory Immunology, Signal Transduction, Stem Cell Markers | |
| 920 | |
| Human, Mouse, Rat, Pig, Canine, Rabbit | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction