missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
CD4 Monoclonal antibody specifically detects CD4 in Ferret samples. It is validated for ELISA
Specifications
Specifications
| Host Species | Rabbit |
| Immunogen | This antibody was obtained from a rabbit immunized with purified, recombinant Ferret CD4 (Accession#: ABS50090.1; Met1-Leu401). |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?