missing translation for 'onlineSavingsMsg'
Learn More

CD4 Antibody (108), HRP, Novus Biologicals™

Product Code. 18324741 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18324741 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18324741 Supplier Novus Biologicals Supplier No. NBP290819H

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

CD4 Monoclonal specifically detects CD4 in Ferret samples. It is validated for ELISA.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD4
Applications ELISA
Classification Monoclonal
Clone 108
Conjugate HRP
Formulation PBS
Gene Alias CD4 antigen, CD4 antigen (p55), CD4 molecule, CD4 receptor, CD4mut, T-cell surface antigen T4/Leu-3, T-cell surface glycoprotein CD4
Host Species Rabbit
Immunogen This antibody was obtained from a rabbit immunized with purified, recombinant Ferret CD4 (ABS50090.1; Met1-Leu401).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Adaptive Immunity, Cell Biology, Cellular Markers, Cytokine Research, Immunology, Innate Immunity, Regulatory Immunology, Signal Transduction, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 920
Target Species Ferret
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.