missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ CD3g Recombinant Rabbit Monoclonal Antibody (K01_1P24)

Product Code. 30826201
Change view
Click to view available options
Quantity:
50 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30826201 100 μL
30828939 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30826201 Supplier Invitrogen™ Supplier No. MA563400

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Recombinant Monoclonal Antibody

CD3g Recombinant Monoclonal Antibody for IHC (P), Western Blot

CD3G is the CD3-gamma polypeptide, which together with CD3-epsilon, -delta and -zeta, and the T-cell receptor alpha/beta and gamma/delta heterodimers, forms the T-cell receptor-CD3 complex. This complex plays an important role in coupling antigen recognition to several intracellular signal-transduction pathways. The genes encoding the epsilon, gamma and delta polypeptides are located in the same cluster on chromosome 11. Defects in the CD3G gene are associated with T cell immunodeficiency.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD3g
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Recombinant Monoclonal
Clone K01_1P24
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation 50mM tris glycine with 40% glycerol, 0.05% BSA, 150mM NaCl and 0.01% sodium azide; pH 7.4
Gene CD3G
Gene Accession No. P09693
Gene Alias CD3 antigen, gamma polypeptide; CD3 gamma-chain; CD3 molecule, gamma; CD3 molecule, gamma polypeptide; Cd3g; CD3g antigen, gamma polypeptide (TiT3 complex); CD3g molecule; CD3g molecule, epsilon (CD3-TCR complex); CD3g molecule, gamma (CD3-TCR complex); CD3-GAMMA; Ctg3; Ctg-3; IMD17; T3G; T-cell antigen receptor complex, gamma subunit of T3; T-cell receptor T3 gamma chain; T-cell surface glycoprotein CD3 gamma chain
Gene Symbols CD3G
Host Species Rabbit
Immunogen A synthetic peptide of human CD3G.
Purification Method Protein A/G
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 917
Target Species Human
Content And Storage Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.