missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CD3g Recombinant Rabbit Monoclonal Antibody (7J2L5)

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA542846
This item is not returnable.
View return policy
Description
Positive test controls include: Rat lung, Rat spleen. Immunogen sequence: GMYQCKGSQN KSKPLQVYYR MCQNCIELNA ATISGFLFAE IVSIFVLAVG VYFIAGQDGV RQSRASDKQT LLPNDQLYQP LKDREDDQYS HLQGNQLRRN.
CD3G is the CD3-gamma polypeptide, which together with CD3-epsilon, -delta and -zeta, and the T-cell receptor alpha/beta and gamma/delta heterodimers, forms the T-cell receptor-CD3 complex. This complex plays an important role in coupling antigen recognition to several intracellular signal-transduction pathways. The genes encoding the epsilon, gamma and delta polypeptides are located in the same cluster on chromosome 11. Defects in the CD3G gene are associated with T cell immunodeficiency.
Specifications
| CD3g | |
| Recombinant Monoclonal | |
| 0.78 mg/mL | |
| PBS with 0.05% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P09693, Q64159 | |
| CD3G | |
| Synthetic peptide. | |
| 100 μL | |
| Primary | |
| Human, Rat | |
| Antibody | |
| IgG |
| ELISA, Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 7J2L5 | |
| Unconjugated | |
| CD3G | |
| CD3 antigen, gamma polypeptide; CD3 gamma-chain; CD3 molecule, gamma; CD3 molecule, gamma polypeptide; Cd3g; CD3g antigen, gamma polypeptide (TiT3 complex); CD3g molecule; CD3g molecule, epsilon (CD3-TCR complex); CD3g molecule, gamma (CD3-TCR complex); CD3-GAMMA; Ctg3; Ctg-3; IMD17; T3G; T-cell antigen receptor complex, gamma subunit of T3; T-cell receptor T3 gamma chain; T-cell surface glycoprotein CD3 gamma chain | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 300678, 917 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction