missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CD3g Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA597562
This item is not returnable.
View return policy
Description
CD3g Polyclonal Antibody for Western Blot, ICC/IF, ELISA
CD3G is the CD3-gamma polypeptide, which together with CD3-epsilon, -delta and -zeta, and the T-cell receptor alpha/beta and gamma/delta heterodimers, forms the T-cell receptor-CD3 complex. This complex plays an important role in coupling antigen recognition to several intracellular signal-transduction pathways. The genes encoding the epsilon, gamma and delta polypeptides are located in the same cluster on chromosome 11. Defects in the CD3G gene are associated with T cell immunodeficiency.
Specifications
| CD3g | |
| Polyclonal | |
| Unconjugated | |
| CD3G | |
| CD3 antigen, gamma polypeptide; CD3 gamma-chain; CD3 molecule, gamma; CD3 molecule, gamma polypeptide; Cd3g; CD3g antigen, gamma polypeptide (TiT3 complex); CD3g molecule; CD3g molecule, epsilon (CD3-TCR complex); CD3g molecule, gamma (CD3-TCR complex); CD3-GAMMA; Ctg3; Ctg-3; IMD17; T3G; T-cell antigen receptor complex, gamma subunit of T3; T-cell receptor T3 gamma chain; T-cell surface glycoprotein CD3 gamma chain | |
| Rabbit | |
| Protein G | |
| RUO | |
| 12502, 300678, 917 | |
| -20°C or -80°C if preferred | |
| Liquid |
| ELISA, Western Blot, Immunocytochemistry | |
| 2.62 mg/mL | |
| PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 | |
| P09693, P11942, Q64159 | |
| CD3G | |
| Recombinant Human T-cell surface glycoprotein CD3 gamma chain protein (138-182AA). | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction