missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CD3g Monoclonal Antibody (D13), Invitrogen™

Mouse Monoclonal Antibody
£161.00 - £304.00
Specifications
| Antigen | CD3g |
|---|---|
| Clone | D13 |
| Concentration | 1 mg/mL |
| Applications | Western Blot |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
17919662
![]() |
Invitrogen™
MA551163 |
200 μL |
£304.00
200µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
17909832
|
Invitrogen™
MA551164 |
20 μL |
£161.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CD3G is the CD3-gamma polypeptide, which together with CD3-epsilon, -delta and -zeta, and the T-cell receptor alpha/beta and gamma/delta heterodimers, forms the T-cell receptor-CD3 complex. This complex plays an important role in coupling antigen recognition to several intracellular signal-transduction pathways. The genes encoding the epsilon, gamma and delta polypeptides are located in the same cluster on chromosome 11. Defects in the CD3G gene are associated with T cell immunodeficiency.Specifications
| CD3g | |
| 1 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.05% Proclin 300; pH 7.4 | |
| CD3 antigen, gamma polypeptide; CD3 gamma-chain; CD3 molecule, gamma; CD3 molecule, gamma polypeptide; Cd3g; CD3g antigen, gamma polypeptide (TiT3 complex); CD3g molecule; CD3g molecule, epsilon (CD3-TCR complex); CD3g molecule, gamma (CD3-TCR complex); CD3-GAMMA; Ctg3; Ctg-3; IMD17; T3G; T-cell antigen receptor complex, gamma subunit of T3; T-cell receptor T3 gamma chain; T-cell surface glycoprotein CD3 gamma chain | |
| CD3G | |
| IgG1 κ | |
| Protein A/G | |
| CD3G |
| D13 | |
| Western Blot | |
| Unconjugated | |
| Mouse | |
| Human | |
| P09693 | |
| 917 | |
| Recombinant Human CD3g protein, Gln23-Asn182 (Accession #P09693). | |
| Primary | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title