missing translation for 'onlineSavingsMsg'
Learn More

CD3e Hamster anti-Mouse, PE-Cy7, Clone: 145-2C11, BD

Hamster Monoclonal Antibody

£62.38 - £174.20

Specifications

Antigen CD3e
Clone 145-2C11
Concentration 0.2mg/mL
Applications Flow Cytometry
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15870769
View Documents
BD Biosciences
561100
25 μg
ONLINE EXCLUSIVE PRICE
£64.20
£62.38 / Each
Save £1.82
2% Off
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
11377823
View Documents
BD Biosciences
552774
0.1 mg
ONLINE EXCLUSIVE PRICE
£182.00
£174.20 / Each
Save £7.80
4% Off
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

The 145-2C11 monoclonal antibody specifically binds to the 25-kDa ε chain of the T-cell receptor-associated CD3 complex that is expressed on thymocytes, mature T lymphocytes, and NK-T cells. The cytoplasmic domain of CD3e participates in the signal transduction events that activate several cellular biochemical pathways as a result of antigen recognition. Soluble 145-2C11 antibody can activate either unprimed (naive) or primed (memory/preactivated) T cells in vivo or in vitro, in the presence of Fc receptor-bearing accessory cells. In contrast, plate-bound 145-2C11 can activate T cells in the absence of accessory cells. Soluble 145-2C11 antibody has been reported to induce re-directed lysis of Fc receptor-bearing target cells by CTL clones and can also block lysis of specific target cells by antigen-specific CTL's. Under some conditions, T-cell activation by 145-2C11 antibody has been reported to result in apoptotic cell death. The 145-2C11 antibody does not cross-react with rat leukocytes. Preincubation of thymus cell suspensions at 37°C for 2-4 hours prior to staining reportedly enhances the ability of anti-CD3ε and anti-αβ TCR mAbs to detect the T-cell receptor on immature thymocytes.

Specifications

Specifications

CD3e
0.2mg/mL
Monoclonal
Hamster
Mouse
H-2Kb specific cytotoxic T lymphocyte clone BM10-37
Primary
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
145-2C11
Flow Cytometry
PE-Cyanine7
RUO
Aqueous buffered solution containing ≤0.09% sodium azide.
IgG1 κ
Affinity Purified
CD3; CD3 epsilon; Cd3e; CD3ε; T3e
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.