missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CD3d Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA588511
This item is not returnable.
View return policy
Description
Immunogen sequence: FKIPIEELED RVFVNCNTSI TWVEGTVGTL LSDITRLDLG KRILDPRGIY RCNGTDIYKD KESTVQVHYR MCQSCVELDP ATVA; Positive Samples: Jurkat; Cellular Location: Membrane, Single-pass type I membrane protein.
CD3d is part of the T-cell receptor/CD3 complex (TCR/CD3 complex) and is involved in T-cell development and signal transduction. The CD3d protein represents the delta subunit of the CD3 complex, and along with four other CD3 subunits, binds either TCR alpha/beta or TCR gamma/delta to form the TCR/CD3 complex on the surface of T-cells. Defects in the CD3d gene are a cause of severe combined immunodeficiency autosomal recessive T-cell-negative/B-cell-positive/NK-cell-positive (SCIDBNK). Two transcript variants encoding different isoforms of CD3d have been found. Other variants of CD3d may also exist, but the full-length nature of their transcripts have yet to be defined.
Specifications
| CD3d | |
| Polyclonal | |
| Unconjugated | |
| Cd3d | |
| CD3 antigen delta polypeptide; CD3 antigen, delta polypeptide; CD3 antigen, delta subunit; CD3 delta; CD3 molecule delta polypeptide; CD3 molecule, delta; CD3D; CD3d antigen, delta polypeptide (TiT3 complex); CD3d molecule; CD3d molecule, delta (CD3-TCR complex); CD3-DELTA; IMD19; OKT3, delta chain; T3D; T-cell receptor T3 delta chain; T-cell surface glycoprotein CD3 delta chain | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 25710, 915 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.27 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P04234, P19377 | |
| Cd3d | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 22-105 of human CD3D (NP_0007231). | |
| 100 μL | |
| Primary | |
| Human, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction