missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ CD39L4/ENTPD5 Antibody Pair [HRP]
Shop All Bio Techne Products
Click to view available options
Quantity:
15 Plates
5 Plates
Unit Size:
Pack of 15
Pack of 5
Description
This solid phase sandwich ELISA utilizes a monoclonal antibody specific for Human CD39L4/ENTPD5.
Specifications
Specifications
| Assay Range | 23.4 pg/mL |
| Assay Sensitivity | 23.4-1500 pg/mL |
| Assay | Sandwich ELISA |
| Conjugate | HRP |
| Product Type | Antibody Pairs |
| Specificity | This solid phase sandwich ELISA utilizes a monoclonal antibody specific for Human CD39L4/ENTPD5. |
| Target Species | Human |
| For Use With (Application) | Sandwich ELISA |
| Gene Alias | CD39 antigen-like 4, CD39L4NTPDase 5, CD39-like 4, EC 3.6.1, EC 3.6.1.42, EC 3.6.1.6, ectonucleoside triphosphate diphosphohydrolase 5, ER-UDPase, GDPase ENTPD5, Guanosine-diphosphatase ENTPD5, MGC163357, MGC163359, NTPDase-5, Nucleoside diphosphatase, PCPH, Pcph proto-oncogene protein, proto-oncogene CPH, UDPase ENTPD5, Uridine-diphosphatase ENTPD5 |
| Gene ID (Entrez) | 957 |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction