missing translation for 'onlineSavingsMsg'
Learn More

CD39L4/ENTPD5 Antibody, Alexa Fluor™ 488, Novus Biologicals™

Product Code. 18355622 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18355622 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18355622 Supplier Novus Biologicals Supplier No. NBP299732AF488

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

CD39L4/ENTPD5 Polyclonal specifically detects CD39L4/ENTPD5 in Human samples. It is validated for ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD39L4/ENTPD5
Applications ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Alexa Fluor 488
Dilution ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate
Gene Alias CD39 antigen-like 4, CD39L4NTPDase 5, CD39-like 4, EC 3.6.1, EC 3.6.1.42, EC 3.6.1.6, ectonucleoside triphosphate diphosphohydrolase 5, ER-UDPase, GDPase ENTPD5, Guanosine-diphosphatase ENTPD5, MGC163357, MGC163359, NTPDase-5, Nucleoside diphosphatase, PCPH, Pcph proto-oncogene protein, proto-oncogene CPH, UDPase ENTPD5, Uridine-diphosphatase ENTPD5
Host Species Rabbit
Immunogen Produced in rabbits immunized with purified, recombinant CD39L4/ENTPD5 (O75356; Met1-His428).
Purification Method Affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Lipid and Metabolism
Primary or Secondary Primary
Gene ID (Entrez) 957
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.