missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD39L3/ENTPD3 Rabbit anti-Human, Clone: 101, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-89707-50UL
This item is not returnable.
View return policy
Description
CD39L3/ENTPD3 Monoclonal specifically detects CD39L3/ENTPD3 in Human samples. It is validated for ELISA.
Specifications
| CD39L3/ENTPD3 | |
| Monoclonal | |
| Unconjugated | |
| 0.2 um filtered solution in PBS | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| ELISA | |
| 101 | |
| ELISA 1:5000-1:10000 | |
| CD39 antigen-like 3, CD39L3EC 3.6.1.5, CD39-like 3, EC 3.6.1, Ecto-apyrase 3, Ecto-ATP diphosphohydrolase 3, Ecto-ATPase 3, Ecto-ATPDase 3, ectonucleoside triphosphate diphosphohydrolase 3, FLJ93839, HB6NTPDase 3, NTPDase-3 | |
| Recombinant Human CD39L3/ENTPD3 protein | |
| 50 μg | |
| Lipid and Metabolism | |
| 956 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction