missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
CD39L3/ENTPD3 Monoclonal antibody specifically detects CD39L3/ENTPD3 in Human samples. It is validated for ELISA
Specifications
Specifications
| Antigen | CD39L3/ENTPD3 |
| Applications | ELISA |
| Classification | Monoclonal |
| Clone | 101 |
| Conjugate | Alexa Fluor 750 |
| Formulation | 50mM Sodium Borate |
| Gene Alias | CD39 antigen-like 3, CD39L3EC 3.6.1.5, CD39-like 3, EC 3.6.1, Ecto-apyrase 3, Ecto-ATP diphosphohydrolase 3, Ecto-ATPase 3, Ecto-ATPDase 3, ectonucleoside triphosphate diphosphohydrolase 3, FLJ93839, HB6NTPDase 3, NTPDase-3 |
| Host Species | Rabbit |
| Immunogen | This antibody was obtained from a rabbit immunized with purified, recombinant Human CD39L3/ENTPD3 (O75355; Gln44-Pro485). |
| Purification Method | Protein A purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?