missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD39L2/ENTPD6 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-58760-25ul
This item is not returnable.
View return policy
Description
CD39L2/ENTPD6 Polyclonal specifically detects CD39L2/ENTPD6 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.
Specifications
| CD39L2/ENTPD6 | |
| Polyclonal | |
| Immunocytochemistry/Immunofluorescence 1-4 μg/mL | |
| CD39 antigen-like 2, CD39L2NTPDase-6, CD39-like 2, dJ738P15.3, DKFZp781G2277, DKFZp781K21102, EC 3.6.1.6, ectonucleoside triphosphate diphosphohydrolase 6, ectonucleoside triphosphate diphosphohydrolase 6 (putative function), ectonucleoside triphosphate diphosphohydrolase 6 (putative), FLJ36711, IL-6SAG, IL6ST2, interleukin 6 signal transducer-2, NTPDase 6 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. |
| Immunocytochemistry, Immunofluorescence | |
| Unconjugated | |
| PBS (pH 7.2), 40% Glycerol with 0.02% Sodium Azide | |
| ENTPD6 | |
| This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:FFSITRAAPGARWGQQAHSPLGTAADGHEVFYGIMFDAGSTGTRVHVFQFTRPPRETPTLTHETFKALKPGLSAYADDVEKSAQGIRELLDVAKQDIPFDFWKATPLVLK | |
| 25 μL | |
| Lipid and Metabolism | |
| 955 | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction