missing translation for 'onlineSavingsMsg'
Learn More

CD39L2/ENTPD6 Antibody (2D10), Novus Biologicals™

Product Code. 18394837 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.01mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18394837 0.1 mg 0.01mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18394837 Supplier Novus Biologicals Supplier No. H00000955M03

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

CD39L2/ENTPD6 Monoclonal antibody specifically detects CD39L2/ENTPD6 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD39L2/ENTPD6
Applications Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 2D10
Conjugate Unconjugated
Dilution Western Blot 1:500, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation In 1x PBS, pH 7.4
Gene Accession No. NP_001238
Gene Alias CD39 antigen-like 2, CD39L2NTPDase-6, CD39-like 2, dJ738P15.3, DKFZp781G2277, DKFZp781K21102, EC 3.6.1.6, ectonucleoside triphosphate diphosphohydrolase 6, ectonucleoside triphosphate diphosphohydrolase 6 (putative function), ectonucleoside triphosphate diphosphohydrolase 6 (putative), FLJ36711, IL-6SAG, IL6ST2, interleukin 6 signal transducer-2, NTPDase 6
Host Species Mouse
Immunogen ENTPD6 (NP_001238, 386 a.a. ~ 484 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. YDLAAGVGLIDAEKGGSLVVGDFEIAAKYVCRTLETQPQSSPFSCMDLTYVSLLLQEFGFPRSKVLKLTRKIDNVETSWALGAIFHYIDSLNRQKSPAS
Purification Method IgG purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Lipid and Metabolism
Primary or Secondary Primary
Gene ID (Entrez) 955
Target Species Human
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.