missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
CD39L1/ENTPD2 Polyclonal antibody specifically detects CD39L1/ENTPD2 in Human samples. It is validated for Immunoprecipitation,Western Blot
Specifications
Specifications
| Antigen | CD39L1/ENTPD2 |
| Applications | Immunoprecipitation, Western Blot |
| Classification | Polyclonal |
| Conjugate | DyLight 755 |
| Formulation | 50mM Sodium Borate |
| Gene Alias | CD39 antigen-like 1, CD39L1NTPDase 2, CD39-like-1, EC 3.6.1, EC 3.6.1.-, EC 3.6.1.5, Ecto-ATP diphosphohydrolase 2, Ecto-ATPase 2, Ecto-ATPDase 2, ectonucleoside triphosphate diphosphohydrolase 2, NTPDase-2 |
| Host Species | Sheep |
| Immunogen | Chinese hamster ovary cell line CHO-derived recombinant human CD39L1/ENTPD2, Thr29-Asp460, Accession # Q9Y5L3 |
| Purification Method | Antigen Affinity-purified |
| Quantity | 0.1 mL |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?