missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD39 Mouse anti-Human, BV421, Clone: TU66, BD Horizon™
Mouse Monoclonal Antibody
Brand: BD Biosciences 566267
This item is not returnable.
View return policy
Description
CD39 Monoclonal antibody specifically detects CD39 in Human samples. It is validated for Flow cytometry.
Specifications
| CD39 | |
| Monoclonal | |
| Brilliant Violet 421 | |
| Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
| Mouse | |
| RUO | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated with BD Horizon™ BV421 under optimum conditions and unconjugated antibody and free BD Horizon™ BV421 were removed. | |
| IgG2b κ |
| Flow Cytometry | |
| TU66 | |
| Vol. Per test: 5μL | |
| ENTPD1; NTPDase-1; Ecto-ATPase 1; Ecto-ATPDase 1 | |
| 25 Tests | |
| Cell Biology | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Reseach Use Only.
Spot an opportunity for improvement?Share a Content Correction