missing translation for 'onlineSavingsMsg'
Learn More

CD39/ENTPD1 Antibody (AC2), Novus Biologicals™

Mouse Monoclonal Antibody

£169.00 - £328.00

Specifications

Antigen CD39/ENTPD1
Clone AC2
Concentration 1 mg/mL
Dilution Western Blot 0.2 μg/mL, Flow Cytometry 1 μg/505 cells, Immunocytochemistry/ Immunofluorescence 1 - 2 μg/mL, Immunoprecipitation 1:10 - 1:500, CyTOF-ready
Applications Western Blot, Flow Cytometry, Immunofluorescence, Immunoprecipitation, CyTOF
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18663109
View Documents
Novus Biologicals
NBP2-50387-0.025ml
0.025 mL
£169.00
0.03mL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18685448
View Documents
Novus Biologicals
NBP2-50387
0.1 mL
£328.00
0.10mL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

CD39/ENTPD1 Monoclonal antibody specifically detects CD39/ENTPD1 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation, CyTOF
Specifications

Specifications

CD39/ENTPD1
1 mg/mL
Western Blot, Flow Cytometry, Immunofluorescence, Immunoprecipitation, CyTOF
Unconjugated
Mouse
Lipid and Metabolism, Myeloid derived Suppressor Cell
PBS
953
IgG1
Protein A purified
AC2
Western Blot 0.2 μg/mL, Flow Cytometry 1 μg/505 cells, Immunocytochemistry/ Immunofluorescence 1 - 2 μg/mL, Immunoprecipitation 1:10 - 1:500, CyTOF-ready
Monoclonal
Purified
RUO
Human
ATPDase, CD39 antigen, CD39EC 3.6.1.5, DKFZp686D194, DKFZp686I093, EC 3.6.1, Ecto-apyrase, Ecto-ATP diphosphohydrolase 1, Ecto-ATPase 1, Ecto-ATPDase 1, ectonucleoside triphosphate diphosphohydrolase 1, FLJ40921, FLJ40959, Lymphoid cell activation antigen, NTPDase 1, NTPDase-1
EBV-transformed human B lymphoblastoid cell line
Primary
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.