missing translation for 'onlineSavingsMsg'
Learn More

CD39/ENTPD1 Antibody (17B5F11), Janelia Fluor™ 549, Novus Biologicals™

Product Code. 18417725 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18417725 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18417725 Supplier Novus Biologicals Supplier No. NBP225223JF549

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

CD39/ENTPD1 Monoclonal antibody specifically detects CD39/ENTPD1 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD39/ENTPD1
Applications Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 17B5F11
Conjugate Janelia Fluor 549
Formulation 50mM Sodium Borate
Gene Alias ATPDase, CD39 antigen, CD39EC 3.6.1.5, DKFZp686D194, DKFZp686I093, EC 3.6.1, Ecto-apyrase, Ecto-ATP diphosphohydrolase 1, Ecto-ATPase 1, Ecto-ATPDase 1, ectonucleoside triphosphate diphosphohydrolase 1, FLJ40921, FLJ40959, Lymphoid cell activation antigen, NTPDase 1, NTPDase-1
Host Species Mouse
Immunogen Amino acids 102-208 of human CD39 were used as the immunogen for the antibody.
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Lipid and Metabolism, Myeloid derived Suppressor Cell
Primary or Secondary Primary
Gene ID (Entrez) 953
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2b λ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.