missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
CD38 Monoclonal specifically detects CD38 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence.
Specifications
Specifications
| Antigen | CD38 |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | AT13/5 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Dilution | Flow Cytometry |
| Formulation | PBS |
| Gene Alias | ADP-ribosyl cyclase 1, ADP-ribosyl cyclase/cyclic ADP-ribose hydrolase, cADPr hydrolase 1, CD38 antigen, CD38 antigen (p45), CD38 molecule, Cyclic ADP-ribose hydrolase 1, EC 3.2.2.5, NAD(+) nucleosidase, T10 |
| Gene Symbols | CD38 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?