missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD38 Mouse, anti-Human, PE, Clone: HIT2, STEMCELL Technologies
Mouse Monoclonal Antibody
£176.85 - £407.70
Specifications
| Antigen | CD38 |
|---|---|
| Clone | HIT2 |
| Applications | Immunohistochemistry, Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence |
| Classification | Monoclonal |
| Conjugate | PE |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
17118451
|
Stemcell Technologies
60014PE.1 |
25 Tests |
£176.85
25 tests |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17108451
|
Stemcell Technologies
60014PE |
100 Tests |
£407.70
100 tests |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
- The HIT2 antibody reacts with CD38, an ∼45 kDa type II transmembrane glycoprotein expressed by many cell types, especially leukocytes
- CD38 is an ectoenzyme (ADP-ribosyl hydrolase) with both cyclase and hydrolase activities, whereby it mediates lymphocyte activation, adhesion and metabolism
- CD31 is a known ligand for the receptor
- In humans, CD38 is expressed by the majority of hematopoietic cells at levels which vary according to the differentiation and activation status of the cells
- It is highly expressed on activated T and B cells and plasma cells, and is also found at high levels in several tissues, including brain, kidney, liver, ovary, pancreas, placenta, testis, and some types of cancerous tissue
- This antibody clone has been verified for purity assessments of cells isolated with EasySep™ kits, including EasySep™ Human CD138 Positive Selection Kit II (Catalog #1877) and EasySep™ Human Whole Blood and Bone Marrow CD138 Positive Selection Kit II (Catalog #17887)
Specifications
| CD38 | |
| Immunohistochemistry, Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence | |
| PE | |
| Chimpanzee, Human | |
| Primary |
| HIT2 | |
| Monoclonal | |
| Mouse | |
| IgG1 κ |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title