missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD38 Mouse, anti-Human, FITC, Clone: AT-1, STEMCELL Technologies
Mouse Monoclonal Antibody
Brand: Stemcell Technologies 60131FI
This item is not returnable.
View return policy
Description
- The AT-1 antibody reacts with CD38, an ∼45 kDa type II integral membrane glycoprotein that is part of the ADP-ribosyl cyclase family
- CD38 is widely expressed at variable levels on hematopoietic cells including early B and T cell lineages, activated B and T cells, NK cells, monocytes, and dendritic cells
- It is strongly expressed in plasma cells
- CD38 is also expressed in some non-hematopoietic tissues such as brain, kidney, muscle, and pancreas
- CD38 is an ectoenzyme that acts as both an ADP-ribosyl cyclase and ADP-ribose (ADPR) hydrolase for the synthesis and hydrolysis of cyclic ADPR (cADPR) and NAADP
- cADPR acts as a second messenger for intracellular calcium mobilization to facilitate glucose-induced insulin secretion
- It has been shown that the AT-1 antibody mutually competes for binding with OKT10 and SUN-4B7, and all three clones only bind the native (not reduced) form of CD38
Specifications
| CD38 | |
| Monoclonal | |
| FITC | |
| 100 Tests | |
| Cynomolgus, Human, Rhesus Monkey | |
| B Cells, Hematopoietic Stem and Progenitor Cells, Plasma |
| Flow Cytometry | |
| AT-1 | |
| Mouse | |
| Primary | |
| IgG1 κ |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction