Learn More
CD38 Mouse anti-Human, Clone: GR7A4 (PE), Abnova™
Mouse Monoclonal Antibody
Brand:  Abnova MAB13886.100Reactions
Product Code. 16092426
  • £349.00 / 100 reactions
Description
Mouse monoclonal antibody raised against human CD38.
  • High specificity
  • High reactivity
Specifications
Specifications
CD38
Monoclonal
PE
Flow Cytometry (20 uL/106 cells) The optimal working dilution should be determined by the end user.
P28907
CD38
Human acute lymphoblastic leukemia (ALL) B cells.
100 Reactions
Primary
Human
Liquid
Flow Cytometry
GR7A4(LD38)
Mouse monoclonal antibody raised against human CD38.
In PBS, pH 7.4 (protein stabilizer, 0.09% sodium azide).
T10
Mouse
Protein A/G purification
RUO
952
Store in the dark at 4°C. Avoid prolonged exposure to light.
IgG1 κ
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
For Research Use Only (RUO)