missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CD38 Monoclonal Antibody (HIT2), Brilliant Violet™ 480, eBioscience™, Invitrogen™

Mouse Monoclonal Antibody
£153.00 - £321.00
Specifications
| Antigen | CD38 |
|---|---|
| Clone | HIT2 |
| Concentration | 5 μL/Test |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
30245075
![]() |
Invitrogen™
414-0389-42 |
100 Tests |
£321.00
Pack of 100 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30245095
|
Invitrogen™
414-0389-41 |
25 Tests |
£153.00
Pack of 25 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CD38 Monoclonal antibody specifically detects CD38 in Human samples. It is validated for Flow CytometrySpecifications
| CD38 | |
| 5 μL/Test | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with BSA and 0.09% sodium azide; pH 7.2 | |
| 2'-phospho-ADP-ribosyl cyclase; 2'-phospho-ADP-ribosyl cyclase/2'-phospho-cyclic-ADP-ribose transferase; 2'-phospho-cyclic-ADP-ribose transferase; ADPRC 1; ADPRC1; ADP-ribosyl cyclase 1; ADP-ribosyl cyclase/cyclic ADP-ribose hydrolase 1; cADPr hydrolase 1; Cd38; CD38 antigen; CD38 antigen (ADP-ribosyl cyclase / cyclic ADP-ribose hydrolase); CD38 antigen (p45); CD38 antigen p45; CD38 molecule; CD38H; Cd38-rs1; cluster of differentiation 38; cyclic ADP-ribose hydrolase 1; ecto-nicotinamide adenine dinucleotide glycohydrolase; I-19; NAD(+) nucleosidase; NAD+ nucleosidase; NIM-R5 antigen; T10 | |
| IgG1 κ | |
| Affinity chromatography | |
| Antibody |
| HIT2 | |
| Flow Cytometry | |
| Brilliant Violet 480 | |
| Mouse | |
| Human | |
| P28907 | |
| 952 | |
| Primary | |
| 4°C, store in dark, DO NOT FREEZE! | |
| CD38 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title