missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD38 Antibody (FS02), Novus Biologicals™
Mouse Monoclonal Antibody
£213.00 - £375.00
Specifications
| Antigen | CD38 |
|---|---|
| Clone | FS02 |
| Concentration | 0.2 mg/mL |
| Dilution | Flow Cytometry 0.5 - 1 ug/million cells in 0.1 ml, SDS-Page, Immunofluorescence 0.5 - 1.0 ug/ml |
| Applications | Flow Cytometry, SDS-Page, Immunofluorescence |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18110948
|
Novus Biologicals
NBP2-44565-0.02MG |
0.02 mg |
£213.00
0.02mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18108758
|
Novus Biologicals
NBP2-44565-0.1MG |
0.1 mg |
£375.00
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Ensure accurate, reproducible results in Flow Cytometry, Immunofluorescence
CD38 Monoclonal specifically detects CD38 in Human samples. It is validated for Flow Cytometry, Immunocytochemistry/Immunofluorescence, Immunofluorescence.Specifications
| CD38 | |
| 0.2 mg/mL | |
| Flow Cytometry, SDS-Page, Immunofluorescence | |
| Unconjugated | |
| Mouse | |
| Apoptosis, Cell Biology, Hematopoietic Stem Cell Markers, Immunology, Stem Cell Markers | |
| 10mM PBS and 0.05% BSA with 0.05% Sodium Azide | |
| 952 | |
| Human CD38 | |
| Primary | |
| Store at 4C. |
| FS02 | |
| Flow Cytometry 0.5 - 1 ug/million cells in 0.1 ml, SDS-Page, Immunofluorescence 0.5 - 1.0 ug/ml | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| ADP-ribosyl cyclase 1, ADP-ribosyl cyclase/cyclic ADP-ribose hydrolase, cADPr hydrolase 1, CD38 antigen, CD38 antigen (p45), CD38 molecule, Cyclic ADP-ribose hydrolase 1, EC 3.2.2.5, NAD(+) nucleosidase, T10 | |
| CD38 | |
| IgG1 κ | |
| Protein A or G purified | |
| This antibody reacts with a type II membrane glycopeptide with a transmembrane sequence near the NH2-terminus. CD38 is a type II transmembrane glycoprotein that is present on early B- and T-cell lineages and activated B- and T-cells but is absent from most mature resting peripheral lymphocytes. CD38 is also found on thymocytes, pre-B cells, germinal center B-cells, mitogen-activated T-cells, monocytes and Ig-secreting plasma cells. CD38 is expressed on CD34+ cells. The CD34+CD38- population of hematopoietic stems cells defines the most pluripotent cells (e.g. blast colony forming cells). |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title