missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD38 Antibody (AT1), Biotin, Novus Biologicals™
Mouse Monoclonal Antibody
£471.00 - £510.00
Specifications
| Antigen | CD38 |
|---|---|
| Clone | AT1 |
| Applications | Flow Cytometry, Immunofluorescence, Immunofluorescence, CyTOF |
| Classification | Monoclonal |
| Conjugate | Biotin |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18617845
|
Novus Biologicals
NBP2-47908B-100ul |
100 μg |
£510.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18606866
|
Novus Biologicals
NBP2-47908B |
0.1 mL |
£471.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CD38 Monoclonal antibody specifically detects CD38 in Human, Monkey samples. It is validated for Flow Cytometry, Immunocytochemistry/ Immunofluorescence, Immunofluorescence, CyTOFSpecifications
| CD38 | |
| Flow Cytometry, Immunofluorescence, Immunofluorescence, CyTOF | |
| Biotin | |
| Mouse | |
| Apoptosis, B Cell Development and Differentiation Markers, Cancer, Hematopoietic Stem Cell Markers, Immunology, Lipid and Metabolism, Neuroscience, Stem Cell Markers | |
| PBS | |
| 952 | |
| IgG1 κ | |
| Protein A or G purified |
| AT1 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Monkey | |
| 2'-phospho-ADP-ribosyl cyclase, 2'-phospho-ADP-ribosyl cyclase/2'-phospho-cyclic-ADP-ribose transferase (EC:2.4.99.20), 2'-phospho-cyclic-ADP-ribose transferase, ADPRC 1, ADPRC1, ADP-ribosyl cyclase 1, ADP-ribosyl cyclase/cyclic ADP-ribose hydrolase, ADP-ribosyl cyclase/cyclic ADP-ribose hydrolase 1, cADPr hydrolase 1, CD_antigen: CD38, CD38, CD38 antigen, CD38 antigen (p45), CD38 molecule, cluster of differentiation 38, Cyclic ADP-ribose hydrolase 1, EC 2.4.99.20, EC 3.2.2.6, EC:3.2.2.6, NAD(+) nucleosidase, T10 | |
| Human T cell line CCRF-CEM | |
| Primary | |
| Store at 4°C in the dark. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title