missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD38 Antibody (AT1), Alexa Fluor™ 700, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-47908AF700
This item is not returnable.
View return policy
Description
CD38 Monoclonal antibody specifically detects CD38 in Human, Monkey samples. It is validated for Flow Cytometry, Immunocytochemistry/ Immunofluorescence, Immunofluorescence, CyTOF
Specifications
| CD38 | |
| Monoclonal | |
| Alexa Fluor 700 | |
| 2'-phospho-ADP-ribosyl cyclase, 2'-phospho-ADP-ribosyl cyclase/2'-phospho-cyclic-ADP-ribose transferase (EC:2.4.99.20), 2'-phospho-cyclic-ADP-ribose transferase, ADPRC 1, ADPRC1, ADP-ribosyl cyclase 1, ADP-ribosyl cyclase/cyclic ADP-ribose hydrolase, ADP-ribosyl cyclase/cyclic ADP-ribose hydrolase 1, cADPr hydrolase 1, CD_antigen: CD38, CD38, CD38 antigen, CD38 antigen (p45), CD38 molecule, cluster of differentiation 38, Cyclic ADP-ribose hydrolase 1, EC 2.4.99.20, EC 3.2.2.6, EC:3.2.2.6, NAD(+) nucleosidase, T10 | |
| Human T cell line CCRF-CEM | |
| 0.1 mL | |
| Apoptosis, B Cell Development and Differentiation Markers, Cancer, Hematopoietic Stem Cell Markers, Immunology, Lipid and Metabolism, Neuroscience, Stem Cell Markers | |
| 952 | |
| Store at 4°C in the dark. | |
| IgG1 κ |
| Flow Cytometry, Immunofluorescence, Immunofluorescence, CyTOF | |
| AT1 | |
| 50 mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human, Monkey | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction